accessibility menu, dialog, popup

UserName

CAS RN 155148-31-5

ClClClClClClClClHNHNNNHNHNNHNHHHHHHHHH

CAS RN 155148-31-5

IUPAC Name: octahydrogen 1-({4-[(1,4,8,11-tetraazacyclotetradecan-1-yl)methyl]phenyl}methyl)-1,4,8,11-tetraazacyclotetradecane octachloride
Synonyms: plerixafor
Molecular Weight (g/mol): 794.46
Molecular Formula: C28H62Cl8N8
InChi Key: UEUPDYPUTTUXLJ-UHFFFAOYSA-N
SMILES: [H+].[H+].[H+].[H+].[H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].C(N1CCCNCCNCCCNCC1)C1=CC=C(CN2CCCNCCNCCCNCC2)C=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results