CAS RN 155026-49-6
IUPAC Name:
benzyl N-[1-({1-[(1-diazo-2-oxoheptan-3-yl)carbamoyl]-2-methylpropyl}carbamoyl)-2-methylpropyl]carbamate
Synonyms:
benzyl N-[1-({1-[(1-diazo-2-oxoheptan-3-yl)carbamoyl]-2-methylpropyl}carbamoyl)-2-methylpropyl]carbamate
Molecular Weight (g/mol):
487.6
Molecular Formula:
C25H37N5O5
InChi Key:
IIPZPLSBQVJCBO-UHFFFAOYNA-N
SMILES:
CCCCC(NC(=O)C(NC(=O)C(NC(=O)OCC1=CC=CC=C1)C(C)C)C(C)C)C(=O)C=[N+]=[N-]
1
–
1
of
1
results
1
Thermo Scientific Chemicals Z-Val-Val-Nle Diazomethyl Ketone
CAS: 155026-49-6 Molecular Formula: C25H37N5O5 Synonym: C-3890; Z-Val-Val-Nle-DMK