accessibility menu, dialog, popup

UserName

CAS RN 1550-35-2

IUPAC Name: 2,4-difluorobenzaldehyde
Synonyms: 2,4-difluorobenzaldehyde
Molecular Weight (g/mol): 142.11
Molecular Formula: C7H4F2O
InChi Key: WCGPCBACLBHDCI-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=O)C(F)=C1
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)
  • (4)