missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzaldehyde 98.0+%, TCI America™

Catalog No. D28395G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28395G 5 g
D283925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28395G Supplier TCI America Supplier No. D28395G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1550-35-2
Molecular Formula C7H4F2O
Molecular Weight (g/mol) 142.11
MDL Number MFCD00010326
InChI Key WCGPCBACLBHDCI-UHFFFAOYSA-N
Synonym 2,4-difluoro-benzaldehyde, benzaldehyde, 2,4-difluoro, pubchem2903, 2,4difluorobenzaldehyde, acmc-209dbv, 2,4-difluorbenzaldehyde, 2,4 difluorobenzaldehyde, 2.4-difluorobenzaldehyde, 2,4-difluoro benzaldehyde, 2.4-difluoro benzaldehyde
PubChem CID 73770
IUPAC Name 2,4-difluorobenzaldehyde
SMILES FC1=CC=C(C=O)C(F)=C1

Specifications

Color Yellow
Quantity 5 g
UN Number 1989
Formula Weight 142.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Difluorobenzaldehyde