accessibility menu, dialog, popup

UserName

CAS RN 1530-32-1

IUPAC Name: ethyltriphenylphosphanium bromide
Synonyms: ethyl(triphenyl)phosphonium bromide
Molecular Weight (g/mol): 371.26
Molecular Formula: C20H20BrP
InChi Key: JHYNXXDQQHTCHJ-UHFFFAOYSA-M
SMILES: [Br-].CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (3)
  • (3)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)