accessibility menu, dialog, popup

UserName

CAS RN 152121-30-7

IUPAC Name: 4-[4-(4-fluorophenyl)-5-(pyridin-4-yl)-2,3-dihydro-1H-imidazol-2-ylidene]cyclohexa-2,5-dien-1-one
Synonyms: 4-[4-(4-fluorophenyl)-5-(pyridin-4-yl)-1,3-dihydroimidazol-2-ylidene]cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 331.35
Molecular Formula: C20H14FN3O
InChi Key: NJNKPVPFGLGHPA-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C1=C(NC(N1)=C1C=CC(=O)C=C1)C1=CC=NC=C1
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results