accessibility menu, dialog, popup

UserName

CAS RN 15182-92-0

IUPAC Name: 4-[2-(dimethylamino)ethoxy]benzaldehyde
Synonyms: 4-[2-(dimethylamino)ethoxy]benzaldehyde
Molecular Weight (g/mol): 193.25
Molecular Formula: C11H15NO2
InChi Key: CBOKAZFQZOQTOC-UHFFFAOYSA-N
SMILES: CN(C)CCOC1=CC=C(C=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results