missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(Diméthylamino)ethoxy]benzaldéhyde, ≥95%, Thermo Scientific™

Numéro de catalogue. CC51504DA
Modifier l'affichage
Click to view available options
Quantité:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC51504DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC51504DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC51504DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 15182-92-0
Formule moléculaire C11H15NO2
Poids moléculaire (g/mol) 193.246
Numéro MDL MFCD07368978
Clé InChI CBOKAZFQZOQTOC-UHFFFAOYSA-N
Synonyme 4-2-dimethylamino ethoxy benzaldehyde, p-2-dimethylamino ethoxy benzaldehyde, 4-2-dimethylamino-ethoxy-benzaldehyde, pubchem12557, 4-2-dimethylaminoethoxy benzaldehyde, 4-2-dimethylamino-ethoxy benzaldehyde, 4-2-dimethylaminoethyloxy benzaldehyde, 4-n,n-dimethylaminoethoxy benzaldehyde, 4-2-dimethylamino-1-ethoxy benzaldehyde
PubChem CID 84823
Nom de l’IUPAC 4-[2-(diméthylamino)étoxy]benzaldéhyde
SOURIRES CN(C)CCOC1=CC=C(C=C1)C=O

Spécifications

Nom ou substance chimique 4-[2-(Dimethylamino)ethoxy]benzaldehyde
CAS % minimum 94.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 95%
Emballage Bouteille en verre ambrée
Quantité 1 g
Poids de formule 193.24
Pourcentage de pureté 95%
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.