accessibility menu, dialog, popup

UserName

CH3OOFFCl

CAS RN 1514-87-0

IUPAC Name: methyl 2-chloro-2,2-difluoroacetate
Synonyms: methyl 2-chloro-2,2-difluoroacetate
Molecular Weight (g/mol): 144.5
Molecular Formula: C3H3ClF2O2
InChi Key: AWUPLMYXZJKHEG-UHFFFAOYSA-N
SMILES: COC(=O)C(F)(F)Cl
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)