missing translation for 'onlineSavingsMsg'
Learn More

Methyl Chlorodifluoroacetate 97.0+%, TCI America™

Catalog No. C15865G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15865G 5 g
C158625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15865G Supplier TCI America Supplier No. C15865G

Chemical Identifiers

CAS 1514-87-0
Molecular Formula C3H3ClF2O2
Molecular Weight (g/mol) 144.502
MDL Number MFCD00000775
InChI Key AWUPLMYXZJKHEG-UHFFFAOYSA-N
Synonym methyl chlorodifluoroacetate, chlorodifluoroacetic acid methyl ester, acetic acid, chlorodifluoro-, methyl ester, methyl difluorochloroacetic acid, methyl chloro difluoro acetate, acetic acid, 2-chloro-2,2-difluoro-, methyl ester, methyl-2-chloro-2,2-difluoroacetate, chloro-difluoro-acetic acid methyl ester, clcf2coome, clcf2co2me
PubChem CID 73935
IUPAC Name methyl 2-chloro-2,2-difluoroacetate
SMILES COC(=O)C(F)(F)Cl

Specifications

Color Colorless
Boiling Point 82°C
Quantity 5 g
UN Number 2924
Formula Weight 144.50
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Chlorodifluoroacetate