CAS RN 15110-17-5
IUPAC Name:
diethyl (3,3-diethoxypropyl)phosphonate
Synonyms:
diethyl 3,3-diethoxypropylphosphonate
Molecular Weight (g/mol):
268.29
Molecular Formula:
C11H25O5P
InChi Key:
WKPXMGJOGYEGNW-UHFFFAOYSA-N
SMILES:
CCOC(CCP(=O)(OCC)OCC)OCC
1
–
1
of
1
results
1
Promotions Available
Diethyl (3,3-diethoxypropyl)phosphonate, 95%, stabilized with max. 1% w/w Na2CO3
Diethyl (3,3-diethoxypropyl)phosphonate, 95%, stabilized with max. 1% w/w Na2CO3
CAS: 15110-17-5 Molecular Formula: C11H25O5P Molecular Weight (g/mol): 268.29 MDL Number: MFCD00800259 InChI Key: WKPXMGJOGYEGNW-UHFFFAOYSA-N Synonym: diethyl 3,3-diethoxypropyl phosphonate,diethyl 3,3-diethoxypropylphosphonate,phosphonic acid, p-3,3-diethoxypropyl-, diethyl ester,3-diethoxyphosphoryl-1,1-diethoxy-propane,3-diethoxyphosphoryl propanaldehyde diethyl acetal,phosphonic acid,p-3,3-diethoxypropyl-, diethyl ester PubChem CID: 328856 IUPAC Name: 3-diethoxyphosphoryl-1,1-diethoxypropane SMILES: CCOC(CCP(=O)(OCC)OCC)OCC