CAS RN 15051-46-4
CAS RN 15051-46-4
IUPAC Name:
{2,2,6,6-tetramethyl-4-[(2-methylprop-2-enoyl)oxy]piperidin-1-yl}oxidanyl
Synonyms:
2,2,6,6-tetramethyl-4-[(2-methylprop-2-enoyl)oxy]piperidin-1-yloxidanyl
Molecular Weight (g/mol):
240.32
Molecular Formula:
C13H22NO3
InChi Key:
BTWSPOZXDCFMLX-UHFFFAOYSA-N
SMILES:
CC(=C)C(=O)OC1CC(C)(C)N([O])C(C)(C)C1
1
–
1
of
1
results
1
4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical 98.0+%, TCI America™
CAS: 15051-46-4 Molecular Formula: C13H22NO3 Molecular Weight (g/mol): 240.32 MDL Number: MFCD17677357 InChI Key: BTWSPOZXDCFMLX-UHFFFAOYSA-N Synonym: 4-Methacryloyloxy-TEMPO Free Radical, TEMPO Methacrylate, PTMA Monomer PubChem CID: 9859588 IUPAC Name: {2,2,6,6-tetramethyl-4-[(2-methylprop-2-enoyl)oxy]piperidin-1-yl}oxidanyl SMILES: CC(=C)C(=O)OC1CC(C)(C)N([O])C(C)(C)C1