missing translation for 'onlineSavingsMsg'
Learn More

4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical 98.0+%, TCI America™

Catalog No. M15315G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M15315G 5 g
M15311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M15315G Supplier TCI America Supplier No. M15315G
Only null left

Chemical Identifiers

CAS 15051-46-4
Molecular Formula C13H22NO3
Molecular Weight (g/mol) 240.32
MDL Number MFCD17677357
InChI Key BTWSPOZXDCFMLX-UHFFFAOYSA-N
Synonym 4-Methacryloyloxy-TEMPO Free Radical, TEMPO Methacrylate, PTMA Monomer
PubChem CID 9859588
IUPAC Name {2,2,6,6-tetramethyl-4-[(2-methylprop-2-enoyl)oxy]piperidin-1-yl}oxidanyl
SMILES CC(=C)C(=O)OC1CC(C)(C)N([O])C(C)(C)C1

Specifications

Melting Point 89°C
Quantity 5 g
Formula Weight 240.32
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical
TSCA No
Recommended Storage Refrigerator