accessibility menu, dialog, popup

UserName

OOHNOO

CAS RN 150256-42-1

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)benzoate
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}benzoate
Molecular Weight (g/mol): 358.37
Molecular Formula: C22H16NO4
InChi Key: CNAVPEPPAVHHKN-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CC=C1NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)