CAS RN 15020-99-2
IUPAC Name:
dichloropalladium; ethane-1,2-diamine
Synonyms:
ethylenediamine; palladium chloride
Molecular Weight (g/mol):
237.42
Molecular Formula:
C2H8Cl2N2Pd
InChi Key:
CAYKJANQVKIYPJ-UHFFFAOYSA-L
SMILES:
Cl[Pd]Cl.NCCN
1
–
1
of
1
results
1
Dichloro(ethylenediamine)palladium(II), Pd 44.8%
CAS: 15020-99-2 Molecular Formula: C2H8Cl2N2Pd Molecular Weight (g/mol): 237.42 MDL Number: MFCD00044991 InChI Key: CAYKJANQVKIYPJ-UHFFFAOYSA-L Synonym: dichloro 1,2-diaminoethane palladium,dichloro ethylenediamine palladium ii,dichloro 1,2-diaminoethane ;palladium,dichloro ethylenediamine palladate ii,ethylenediamine palladium ii chloride,ethylenediamine; palladium chloride,palladium ii chloride ethylenediamine complex,dichloro ethylenediamine palladium ii , pd PubChem CID: 11139157 SMILES: Cl[Pd]Cl.NCCN