missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloro(ethylenediamine)palladium(II), Pd 44.8%
Description
Catalyst for C-C coupling reactions like Sonogashira coupling. Formation of host-guest complexes. Synthesis of mixed-metal, mixed-pyrimidine self-assembling metallacalix[n]arenes; ring-in-ring complex; binding of metal with glycose phosphates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 1 g |
| Sensitivity | Hygroscopic |
| Solubility Information | Very low solubility in water. |
| IUPAC Name | dichloropalladium; ethane-1,2-diamine |
| Formula Weight | 237.41 |
| Concentration or Composition (by Analyte or Components) | Pd 44.8% |
| Physical Form | Crystalline |
| Chemical Name or Material | Dichloro(ethylenediamine)palladium(II) |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only