CAS RN 1499-10-1
9,10-Diphenylanthracene, 98%
CAS: 1499-10-1 Molecular Formula: C26H18 Molecular Weight (g/mol): 330.43 MDL Number: MFCD00001253 InChI Key: FCNCGHJSNVOIKE-UHFFFAOYSA-N Synonym: anthracene, 9,10-diphenyl,9,10-diphenyl anthracene,unii-51bq8iyq9u,51bq8iyq9u,acmc-1brsc,9, 9-diphenylanthracene,anthracene,10-diphenyl,9,10-diphenylanthracene,9,10-diphenylanthracene 1g,9,10-diphenylanthracene, analytical standard PubChem CID: 15159 ChEBI: CHEBI:51676 IUPAC Name: 9,10-diphenylanthracene SMILES: C1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12
9,10-Diphenylanthracene, 99%
CAS: 1499-10-1 Molecular Formula: C26H18 Molecular Weight (g/mol): 330.43 MDL Number: MFCD00001253 InChI Key: FCNCGHJSNVOIKE-UHFFFAOYSA-N Synonym: anthracene, 9,10-diphenyl,9,10-diphenyl anthracene,unii-51bq8iyq9u,51bq8iyq9u,acmc-1brsc,9, 9-diphenylanthracene,anthracene,10-diphenyl,9,10-diphenylanthracene,9,10-diphenylanthracene 1g,9,10-diphenylanthracene, analytical standard PubChem CID: 15159 ChEBI: CHEBI:51676 IUPAC Name: 9,10-diphenylanthracene SMILES: C1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12
9,10-Diphenylanthracene, 98%, Thermo Scientific™
9,10-Diphenylanthracene 98.0+%, TCI America™
CAS: 1499-10-1 Molecular Formula: C26H18 Molecular Weight (g/mol): 330.43 MDL Number: MFCD00001253 InChI Key: FCNCGHJSNVOIKE-UHFFFAOYSA-N Synonym: anthracene, 9,10-diphenyl,9,10-diphenyl anthracene,unii-51bq8iyq9u,51bq8iyq9u,acmc-1brsc,9, 9-diphenylanthracene,anthracene,10-diphenyl,9,10-diphenylanthracene,9,10-diphenylanthracene 1g,9,10-diphenylanthracene, analytical standard PubChem CID: 15159 ChEBI: CHEBI:51676 IUPAC Name: 9,10-diphenylanthracene SMILES: C1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12
9,10-Diphenylanthracene (purified by sublimation) 99.0+%, TCI America™
CAS: 1499-10-1 Molecular Formula: C26H18 Molecular Weight (g/mol): 330.43 MDL Number: MFCD00001253 InChI Key: FCNCGHJSNVOIKE-UHFFFAOYSA-N Synonym: anthracene, 9,10-diphenyl,9,10-diphenyl anthracene,unii-51bq8iyq9u,51bq8iyq9u,acmc-1brsc,9, 9-diphenylanthracene,anthracene,10-diphenyl,9,10-diphenylanthracene,9,10-diphenylanthracene 1g,9,10-diphenylanthracene, analytical standard PubChem CID: 15159 ChEBI: CHEBI:51676 IUPAC Name: 9,10-diphenylanthracene SMILES: C1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12