missing translation for 'onlineSavingsMsg'
Learn More

9,10-Diphenylanthracene (purified by sublimation) 99.0+%, TCI America™

Catalog No. D44011G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D44011G Supplier TCI America Supplier No. D44011G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1499-10-1
Molecular Formula C26H18
Molecular Weight (g/mol) 330.43
MDL Number MFCD00001253
InChI Key FCNCGHJSNVOIKE-UHFFFAOYSA-N
Synonym anthracene, 9,10-diphenyl, 9,10-diphenyl anthracene, unii-51bq8iyq9u, 51bq8iyq9u, acmc-1brsc, 9, 9-diphenylanthracene, anthracene,10-diphenyl, 9,10-diphenylanthracene, 9,10-diphenylanthracene 1g, 9,10-diphenylanthracene, analytical standard
PubChem CID 15159
ChEBI CHEBI:51676
IUPAC Name 9,10-diphenylanthracene
SMILES C1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12

Specifications

Melting Point 252°C
Color Yellow
Boiling Point 485°C
Quantity 1 g
Formula Weight 330.43
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Diphenylanthracene (purified by sublimation)
EINECSNumber (4)-1873
RTECSNumber CB0545500
TSCA Yes