accessibility menu, dialog, popup

UserName

NaNaOHPOOSClPHOOO

CAS RN 149845-07-8

IUPAC Name: disodium hydrogen {[(4-chlorophenyl)sulfanyl](hydrogen phosphonato)methyl}phosphonate
Synonyms: tiludronic acid
Molecular Weight (g/mol): 362.56
Molecular Formula: C7H7ClNa2O6P2S
InChi Key: SKUHWSDHMJMHIW-UHFFFAOYSA-L
SMILES: [Na+].[Na+].OP([O-])(=O)C(SC1=CC=C(Cl)C=C1)P(O)([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results