missing translation for 'onlineSavingsMsg'
Learn More

Disodium Tiludronate 98.0+%, TCI America™

Catalog No. D416725MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D416725MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D416725MG Supplier TCI America Supplier No. D416725MG
Only null left

Chemical Identifiers

CAS 149845-07-8
Molecular Formula C7H7ClNa2O6P2S
Molecular Weight (g/mol) 362.564
MDL Number MFCD04039691
InChI Key SKUHWSDHMJMHIW-UHFFFAOYSA-L
Synonym Tiludronic Acid Disodium Salt, Disodium [[(4-Chlorophenyl)thio]methylene]bis(hydrogenphosphonate)
PubChem CID 60936
IUPAC Name disodium;[(4-chlorophenyl)sulfanyl-[hydroxy(oxido)phosphoryl]methyl]-hydroxyphosphinate
SMILES C1=CC(=CC=C1SC(P(=O)(O)[O-])P(=O)(O)[O-])Cl.[Na+].[Na+]

Specifications

Color White
Quantity 25 mg
Formula Weight 362.56
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Disodium Tiludronate
TSCA No
Recommended Storage Freezer (-20 °C)