accessibility menu, dialog, popup

UserName

CAS RN 149-57-5

IUPAC Name: 2-ethylhexanoic acid
Synonyms: 2-ethylhexanoic acid tridecyl ethylhexanoate
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: OBETXYAYXDNJHR-UHFFFAOYNA-N
SMILES: CCCCC(CC)C(O)=O
Boiling Point 
  • (4)
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (4)
  • (6)
Percent Purity 
  • (2)
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (2)
  • (8)
  • (4)
  • (2)
  • (4)
  • (4)
  • (6)
  • (2)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)