missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexanoic acid, 99%

Catalog No. AAA12644AP
Change view
Click to view available options
Quantity:
10,000 mL
100 mL
500 mL
2500 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA12644AP 500 mL
AAA12644AE 100 mL
AAA126440F 2500 mL
AAA126440D 10,000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA12644AP Supplier Thermo Scientific Chemicals Supplier No. A12644AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 149-57-5 | C8H16O2 | 144.214 g/mol

2-Ethylhexanoic acid is used in the preparation of metal derivatives, which act as a catalyst in polymerization reactions. For example, tin 2-ethylhexanoate is used in the manufacturing of poly(lactic-co-glycolic acid). It is also used as a stabilizer for polyvinyl chlorides. It is also involved in solvent extraction and dye granulation. Further, it is used to prepare plasticizers, lubricants, detergents, flotation aids, corrosion inhibitors and alkyd resins. In addition to this, it serves as a catalyst for polyurethane foaming.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylhexanoic acid is used in the preparation of metal derivatives, which act as a catalyst in polymerization reactions. For example, tin 2-ethylhexanoate is used in the manufacturing of poly(lactic-co-glycolic acid). It is also used as a stabilizer for polyvinyl chlorides. It is also involved in solvent extraction and dye granulation. Further, it is used to prepare plasticizers, lubricants, detergents, flotation aids, corrosion inhibitors and alkyd resins. In addition to this, it serves as a catalyst for polyurethane foaming.

Solubility
Miscible with water.

Notes
Incompatible with bases, oxidizing agents and reducing agents.

Chemical Identifiers

CAS 149-57-5
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00002675
InChI Key OBETXYAYXDNJHR-UHFFFAOYSA-N
Synonym 2-ethylcaproic acid, hexanoic acid, 2-ethyl, ethylhexanoic acid, ethylhexoic acid, 2-ethylhexoic acid, butylethylacetic acid, 2-butylbutanoic acid, 3-heptanecarboxylic acid, ethyl hexanoic acid, 2-ethyl-hexoic acid
PubChem CID 8697
IUPAC Name 2-ethylhexanoic acid
SMILES CCCCC(CC)C(=O)O

Specifications

Melting Point -59°C
Density 0.91
pH 3
Boiling Point 226°C to 229°C
Flash Point 114°C (237°F)
Refractive Index 1.425
Quantity 500 mL
Beilstein 1750468
Solubility Information Miscible with water.
Formula Weight 144.21
Percent Purity 99%
Chemical Name or Material 2-Ethylhexanoic acid
Show More Show Less
Hazard Category H302-H312-H314-H361d
Hazard Statement GHS H Statement
H361-H302-H312
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P201-P202-P260-P264b-P270-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 205-743-6
RTECSNumber MO7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only