missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylhexanoic acid, ≥99.5% (GC), MilliporeSigma™ Supelco™
Description
Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
Specifications
Specifications
| Chemical Name or Material | 2-Ethylhexanoic Acid |
| CAS | 149-57-5 |
| Ignition Point | 699°F |
| Vapor Pressure | 10 mmHg (115°C) |
| Vapor Density | 4.98 (vs. air) |
| Density | 0.903 g/mL (at 25°C) |
| Flash Point | 114°C - Closed Cup |
| Linear Formula | CH3(CH2)3 CH(C2H5)CO2 H |
| Technique | Gas Chromatography |
| MDL Number | MFCD00002675 |
| Show More |