missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexanoic Acid 99.0+%, TCI America™

Catalog No. E0120500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0120500ML 500 mL
E012025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0120500ML Supplier TCI America Supplier No. E0120500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 149-57-5
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00002675
InChI Key OBETXYAYXDNJHR-UHFFFAOYSA-N
Synonym 2-ethylcaproic acid, hexanoic acid, 2-ethyl, ethylhexanoic acid, ethylhexoic acid, 2-ethylhexoic acid, butylethylacetic acid, 2-butylbutanoic acid, 3-heptanecarboxylic acid, ethyl hexanoic acid, 2-ethyl-hexoic acid
PubChem CID 8697
IUPAC Name 2-ethylhexanoic acid
SMILES CCCCC(CC)C(=O)O

Specifications

Color Colorless
Boiling Point 228°C
Quantity 500 mL
UN Number 3265
Formula Weight 144.21
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexanoic Acid
EINECSNumber (2)-0608
RTECSNumber MO7700000
TSCA Yes