CAS RN 1482416-52-3
IUPAC Nor:
1-bromo-1,1,2,2,4,5,5,7,8,8,10,11,11,13,14,14,16,16,17,17,17-henicosafluoro-4,7,10,13-tetrakis(trifluoromethyl)-3,6,9,12,15-pentaoxaheptadecane
Synonymes:
1-bromo-1,1,2,2,4,5,5,7,8,8,10,11,11,13,14,14,16,16,17,17,17-henicosafluoro-4,7,10,13-tetrakis(trifluoromethyl)-3,6,9,12,15-pentaoxaheptadecane
Poids moléculaire (g/mol):
979.02
Formule moléculaire:
C16BrF33O5
InChi Key:
VAXWUAHPDZRLAP-UHFFFAOYNA-N
SMILES:
FC(F)(F)C(F)(F)OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)Br)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F
1
–
1
de
1
résultats
1
1-Bromoperfluoro-4,7,10,13-tétraméthyl-3,6,9,12,15-pentaoxaheptadécane 94,0+%, TCI America™
CAS: 1482416-52-3 Formule moléculaire: C16BrF33O5 Poids moléculaire (g/mol): 979.02 Numéro MDL: MFCD27918480 Clé InChI: VAXWUAHPDZRLAP-UHFFFAOYNA-N Synonyme: 1-Bromoperfluoro-4,7,10,13-tetramethyl-3,6,9,12,15-pentaoxaheptadecane PubChem CID: 91972086 Nom de l’IUPAC: 1-bromo-1,1,2,2,4,5,5,7,8,8,10,11,11,13,14,14,16,16,17,17,17-henicosafluoro-4,7,10,13-tétrakis(trifluorométhyl)-3,6,9,12,15-pentaoxaheptadécane SOURIRES: FC(F)(F)C(F)(F)OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)Br)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F