missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoperfluoro-4,7,10,13-tetramethyl-3,6,9,12,15-pentaoxaheptadecane 94.0+%, TCI America™

Catalog No. B41675G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41675G 5 g
B41671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41675G Supplier TCI America Supplier No. B41675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1482416-52-3
Molecular Formula C16BrF33O5
Molecular Weight (g/mol) 979.02
MDL Number MFCD27918480
InChI Key VAXWUAHPDZRLAP-UHFFFAOYNA-N
Synonym 1-Bromoperfluoro-4,7,10,13-tetramethyl-3,6,9,12,15-pentaoxaheptadecane
PubChem CID 91972086
IUPAC Name 1-bromo-1,1,2,2,4,5,5,7,8,8,10,11,11,13,14,14,16,16,17,17,17-henicosafluoro-4,7,10,13-tetrakis(trifluoromethyl)-3,6,9,12,15-pentaoxaheptadecane
SMILES FC(F)(F)C(F)(F)OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)Br)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F

Specifications

Quantity 5 g
Formula Weight 979.02
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-1,1,2,2,4,5,5,7,8,8,10,11,11,13,14,14,16,16,17,17,17-henicosafluoro-4,7,10,13-tetrakis(trifl