CAS RN 148-86-7
CAS RN 148-86-7
IUPAC Name:
[1,1'-biphenyl]-4-yl acetate
Synonyms:
[1,1'-biphenyl]-4-yl acetate
Molecular Weight (g/mol):
212.25
Molecular Formula:
C14H12O2
InChi Key:
MISFQCBPASYYGV-UHFFFAOYSA-N
SMILES:
CC(=O)OC1=CC=C(C=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
4-Acetoxybiphenyl 99.0+%, TCI America™
CAS: 148-86-7 Molecular Formula: C14H12O2 Molecular Weight (g/mol): 212.25 MDL Number: MFCD00014979 InChI Key: MISFQCBPASYYGV-UHFFFAOYSA-N Synonym: 4-acetoxybiphenyl,4-biphenylyl acetate,p-phenylphenyl acetate,4-biphenylol, acetate,1,1'-biphenyl-4-yl acetate,acetic acid 4-biphenyl ester,1,1'-biphenyl-4-ol,4-acetate,4-biphenyl acetate,p-phenylphenol acetate,4-phenylphenol acetate PubChem CID: 346066 IUPAC Name: [1,1'-biphenyl]-4-yl acetate SMILES: CC(=O)OC1=CC=C(C=C1)C1=CC=CC=C1