missing translation for 'onlineSavingsMsg'
Learn More

4-Acetoxybiphenyl 99.0+%, TCI America™

Catalog No. A103225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A103225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A103225G Supplier TCI America Supplier No. A103225G
Only null left

Chemical Identifiers

CAS 148-86-7
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00014979
InChI Key MISFQCBPASYYGV-UHFFFAOYSA-N
Synonym 4-acetoxybiphenyl, 4-biphenylyl acetate, p-phenylphenyl acetate, 4-biphenylol, acetate, 1,1'-biphenyl-4-yl acetate, acetic acid 4-biphenyl ester, 1,1'-biphenyl-4-ol,4-acetate, 4-biphenyl acetate, p-phenylphenol acetate, 4-phenylphenol acetate
PubChem CID 346066
IUPAC Name [1,1'-biphenyl]-4-yl acetate
SMILES CC(=O)OC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 87°C
Color White
Quantity 25 g
Formula Weight 212.25
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetoxybiphenyl