missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ 8-Quinolinol, 98-100% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013166
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013166 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013166 Supplier Merck Emd Millipore Supplier No. 36524250MG

Chemical structure: quinolone

Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Product Line PESTANAL
CAS 148-24-3
Physical Form Neat
Linear Formula C9H7 NO
MDL Number MFCD00006807
Melting Point 70°C to 73°C
Boiling Point 267°C (752 mmHg)
Synonym 8-Hydroxyquinoline; 8-Oxychinolin; Oxine
Type Analytical Standard
Quantity 250 mg
Formula Weight 145.16
Percent Purity 98 to 100% (GC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH