missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Hydroxyquinoline, ACS Reagent Grade, Ricca Chemical
Specifications
Specifications
| CAS Min % | 98.0 |
| CAS Max % | 100.0 |
| Packaging | White Poly Wide Mouth Bottle |
| Quantity | 100 g |
| Grade | ACS |
| Chemical Name or Material | 8-Hydroxyquinoline |
Safety and Handling
| ShelfLife | 60 months |