missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline, ACS Grade, LabChem™

Catalog No. LC155759
Change view
Click to view available options
Quantity:
100 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC155759 100 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC155759 Supplier LabChem Supplier No. LC155759

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 148-24-3
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00006807
InChI Key MCJGNVYPOGVAJF-UHFFFAOYSA-N
Synonym 8-hydroxyquinoline, 8-quinolinol, oxyquinoline, oxine, quinophenol, 8-quinol, 8-oxyquinoline, phenopyridine, oxychinolin, bioquin
PubChem CID 1923
ChEBI CHEBI:48981
IUPAC Name quinolin-8-ol
SMILES OC1=C2N=CC=CC2=CC=C1

Specifications

CAS Max % 100
Color White
Boiling Point 267°C
Packaging Amber Glass
Linear Formula HOC6H3N:CHCH:CH
Quantity 100 g
Solubility Information Soluble in water
Formula Weight 145.16
Grade ACS
Physical Form Crystals
Chemical Name or Material 8-Hydroxyquinoline
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Avoid breathing dust.
Do not eat, drink or smoke when using this product.
Wash exposed skin thoroughly after handling.
Store locked up in a well-ventilated place.
Keep container tightly closed.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 205-711-1
Recommended Storage Room Temperature