accessibility menu, dialog, popup

UserName

H3CSiCH3ClCl

CAS RN 14799-94-1

IUPAC Name: dichloro(hexyl)methylsilane
Synonyms: dichloro(hexyl)methylsilane
Molecular Weight (g/mol): 199.19
Molecular Formula: C7H16Cl2Si
InChi Key: KKRMHVJQWMXYBZ-UHFFFAOYSA-N
SMILES: CCCCCC[Si](C)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results