accessibility menu, dialog, popup

UserName

CAS RN 14678-02-5

IUPAC Name: 3-methyl-1,2-oxazol-5-amine
Synonyms: 3-methyl-1,2-oxazol-5-amine
Molecular Weight (g/mol): 98.11
Molecular Formula: C4H6N2O
InChi Key: FNXYWHTZDAVRTB-UHFFFAOYSA-N
SMILES: CC1=NOC(N)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)