accessibility menu, dialog, popup

UserName

H3COOH3COOCH3

CAS RN 1462-12-0

IUPAC Name: 1,3-diethyl 2-ethylidenepropanedioate
Synonyms: 1,3-diethyl 2-ethylidenepropanedioate
Molecular Weight (g/mol): 186.21
Molecular Formula: C9H14O4
InChi Key: LBBAWVLUOZVYCC-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=CC)C(=O)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results