missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ethylidenemalonate, 99%

Catalog No. AAL1211706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1211706 5 g
AAL1211703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1211706 Supplier Thermo Scientific Chemicals Supplier No. L1211706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1462-12-0 | C9H14O4 | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1462-12-0
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.21
MDL Number MFCD00009145
InChI Key LBBAWVLUOZVYCC-UHFFFAOYSA-N
Synonym diethyl ethylidenemalonate, diethyl 2-ethylidenemalonate, propanedioic acid, ethylidene-, diethyl ester, 1,3-diethyl 2-ethylidenepropanedioate, 2-ethylidenemalonic acid diethyl ester, diethylethylidenemalonate, acmc-1c0mi, diethyl ethylidene malonate, diethyl ethylidenepropanedioate, diethyl 2-ethylidenemalonate #
PubChem CID 73831
SMILES CCOC(=O)C(=CC)C(=O)OCC

Specifications

Density 1.019
Boiling Point 115°C to 116°C (17 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.442
Quantity 5 g
Beilstein 1773932
IUPAC Name 1,3-diethyl 2-ethylidenepropanedioate
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material Diethyl ethylidenemalonate
EINECSNumber 215-965-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only