CAS RN 146137-76-0
IUPAC Name:
2-fluoro-5-iodobenzaldehyde
Synonyms:
2-fluoro-5-iodobenzaldehyde
Molecular Weight (g/mol):
250.01
Molecular Formula:
C7H4FIO
InChi Key:
BIRCCQCPGMMGPJ-UHFFFAOYSA-N
SMILES:
FC1=C(C=O)C=C(I)C=C1
1
–
1
of
1
results
1
Promotions Available
2-Fluoro-5-iodobenzaldehyde, 97+%
2-Fluoro-5-iodobenzaldehyde, 97+%
CAS: 146137-76-0 Molecular Formula: C7H4FIO Molecular Weight (g/mol): 250.01 MDL Number: MFCD03093999 InChI Key: BIRCCQCPGMMGPJ-UHFFFAOYSA-N Synonym: 2-fluoro-5-iodo-benzaldehyde,pubchem4222,acmc-1bypb,c7h4flo,6-fluoro-3-iodobenzaldehyde,2-fluoranyl-5-iodanyl-benzaldehyde,2-fluoro-5-iodobenzaldehyde PubChem CID: 16217058 IUPAC Name: 2-fluoro-5-iodobenzaldehyde SMILES: FC1=C(C=O)C=C(I)C=C1