missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-iodobenzaldehyde, 97+%

Catalog No. AAH3203314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3203314 25 g
AAH3203306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3203314 Supplier Thermo Scientific Chemicals Supplier No. H3203314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 146137-76-0 | C7H4FIO | 250.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 146137-76-0
Molecular Formula C7H4FIO
Molecular Weight (g/mol) 250.01
MDL Number MFCD03093999
InChI Key BIRCCQCPGMMGPJ-UHFFFAOYSA-N
Synonym 2-fluoro-5-iodo-benzaldehyde, pubchem4222, acmc-1bypb, c7h4flo, 6-fluoro-3-iodobenzaldehyde, 2-fluoranyl-5-iodanyl-benzaldehyde, 2-fluoro-5-iodobenzaldehyde
PubChem CID 16217058
IUPAC Name 2-fluoro-5-iodobenzaldehyde
SMILES FC1=C(C=O)C=C(I)C=C1

Specifications

Melting Point 42°C to 46°C
Quantity 25 g
Sensitivity Air and light sensitive
Formula Weight 250.01
Percent Purity ≥97%
Chemical Name or Material 2-Fluoro-5-iodobenzaldehyde
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only