CAS RN 144900-57-2
IUPAC Name:
1-(2-chloropyridin-4-yl)methanamine
Synonyms:
1-(2-chloropyridin-4-yl)methanamine
Molecular Weight (g/mol):
142.59
Molecular Formula:
C6H7ClN2
InChi Key:
GGHCWJWUOSNCSK-UHFFFAOYSA-N
SMILES:
NCC1=CC(Cl)=NC=C1
1
–
1
of
1
results
1
4-Aminomethyl-2-chloropyridine, 95%
CAS: 144900-57-2 Molecular Formula: C6H7ClN2 Molecular Weight (g/mol): 142.59 InChI Key: GGHCWJWUOSNCSK-UHFFFAOYSA-N Synonym: 2-chloropyridin-4-yl methanamine,4-pyridinemethanamine, 2-chloro,2-chloropyridin-4-yl methylamine,4-aminomethyl-2-chloropyridine,2-chloro-4-aminomethylpyridine,2-chloro-4-pyridyl methylamine,1-2-chloro-4-pyridinyl methanamine,2-chloro-4-pyridinemethanamine,1-2-chloropyridin-4-yl methanamine,c-2-chloro-pyridin-4-yl-methylamine PubChem CID: 2794167 IUPAC Name: (2-chloropyridin-4-yl)methanamine SMILES: C1=CN=C(C=C1CN)Cl