accessibility menu, dialog, popup

UserName

CH3OHBr

CAS RN 14472-14-1

IUPAC Name: 4-bromo-3-methylphenol
Synonyms: phenol, 4-bromo-3-methyl-
Molecular Weight (g/mol): 187.04
Molecular Formula: C7H7BrO
InChi Key: GPOQODYGMUTOQL-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CC=C1Br
Boiling Point 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)