missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-methylphenol, 98%

Catalog No. AAL1751006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1751006 5 g
AAL1751003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1751006 Supplier Thermo Scientific Chemicals Supplier No. L1751006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14472-14-1 | C7H7BrO | 187.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14472-14-1
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.04
MDL Number MFCD00079723
InChI Key GPOQODYGMUTOQL-UHFFFAOYSA-N
Synonym 4-bromo-3-methyl-phenol, 4-bromo-m-cresol, phenol, 4-bromo-3-methyl, 4-bromo-3-methyl phenol, 4-bromo-5-methyl-phenol, 4-brom-3-methylphenol, zlchem 434, pubchem2201, 3-methyl-4-bromophenol, 2-bromo-5-hydroxytoluene
PubChem CID 72857
IUPAC Name 4-bromo-3-methylphenol
SMILES CC1=CC(O)=CC=C1Br

Specifications

Melting Point 57°C to 61°C
Boiling Point 142°C to 145°C (23 mmHg)
Odor Phenol-like
Quantity 5 g
Beilstein 1859122
Formula Weight 187.04
Percent Purity 98%
Chemical Name or Material 4-Bromo-3-methylphenol
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 238-464-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only