accessibility menu, dialog, popup

UserName

CAS RN 14451-86-6

H3COH

CAS RN 14451-86-6

IUPAC Name: 2-butyladamantan-2-ol
Synonyms: 2-butyladamantan-2-ol
Molecular Weight (g/mol): 208.35
Molecular Formula: C14H24O
InChi Key: PXRAOOXXPGRCJZ-UHFFFAOYSA-N
SMILES: CCCCC1(O)C2CC3CC(C2)CC1C3
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results