missing translation for 'onlineSavingsMsg'
Learn More

2-Butyl-2-adamantanol 98.0+%, TCI America™

Catalog No. B28395G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B28395G Supplier TCI America Supplier No. B28395G
Only null left

Specifications

CAS 14451-86-6
Melting Point 79°C
Color White-Yellow
Molecular Formula C14H24O
Quantity 5 g
Synonym 2-Butyl-2-hydroxyadamantane
InChI Key PXRAOOXXPGRCJZ-UHFFFAOYSA-N
SMILES CCCCC1(C2CC3CC(C2)CC1C3)O
IUPAC Name 2-butyladamantan-2-ol
Molecular Weight (g/mol) 208.345
PubChem CID 12627253
Formula Weight 208.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Butyl-2-adamantanol
Show More Show Less