accessibility menu, dialog, popup

UserName

CAS RN 1445-19-8

IUPAC Name: sodium 8-(phenylamino)naphthalene-1-sulfonate
Synonyms: sodium 8-(phenylamino)naphthalene-1-sulfonate
Molecular Weight (g/mol): 321.33
Molecular Formula: C16H12NNaO3S
InChi Key: VUSNMYYVKMGHEH-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=C2C(NC3=CC=CC=C3)=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results