missing translation for 'onlineSavingsMsg'
Learn More

ANS-Na (=Sodium 8-Anilino-1-naphthalenesulfonate) 97.0+%, TCI America™

Catalog No. A53525G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A53525G 5 g
A53521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A53525G Supplier TCI America Supplier No. A53525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1445-19-8
Molecular Formula C16H12NNaO3S
Molecular Weight (g/mol) 321.33
MDL Number MFCD00054279
InChI Key VUSNMYYVKMGHEH-UHFFFAOYSA-M
Synonym ans-na, sodium 8-anilinonaphthalene-1-sulphonate, 1-naphthalenesulfonic acid, 8-phenylamino-, monosodium salt, 1,8-phenyl acid sodium salt, sodium 8-anilino-1-naphthalenesulfonate, 1-naphthalenesulfonic acid, 8-phenylamino-, sodium salt 1:1, 82-76-8 parent, sodium 8-phenylamino naphthalene-1-sulfonate, 1-naphthalenesulfonic acid, 8-anilino-, monosodium salt, sodium 8-anilinonaphthalene-1-sulfonate
PubChem CID 23670839
IUPAC Name sodium 8-(phenylamino)naphthalene-1-sulfonate
SMILES [Na+].[O-]S(=O)(=O)C1=C2C(NC3=CC=CC=C3)=CC=CC2=CC=C1

Specifications

Quantity 5 g
Formula Weight 321.33
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material ANS-Na (=Sodium 8-Anilino-1-naphthalenesulfonate) [Hydrophobic fluorescent probe]