accessibility menu, dialog, popup

UserName

CAS RN 1440-61-5

ClONO

CAS RN 1440-61-5

IUPAC Name: 2-chloro-1-(morpholin-4-yl)ethan-1-one
Synonyms: morpholine, 4-(chloroacetyl)-
Molecular Weight (g/mol): 163.6
Molecular Formula: C6H10ClNO2
InChi Key: YMQRPXBBBOXHNZ-UHFFFAOYSA-N
SMILES: ClCC(=O)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results