accessibility menu, dialog, popup

UserName

OHBrHO

CAS RN 14381-51-2

IUPAC Name: 3-bromobenzene-1,2-diol
Synonyms: 3-bromobenzene-1,2-diol
Molecular Weight (g/mol): 189.01
Molecular Formula: C6H5BrO2
InChi Key: JPBDMIWPTFDFEU-UHFFFAOYSA-N
SMILES: OC1=CC=CC(Br)=C1O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)