accessibility menu, dialog, popup

UserName

CAS RN 1438-82-0

CH3SiH3COSiCH3CH3H3C

CAS RN 1438-82-0

IUPAC Name: dimethyl[(trimethylsilyl)oxy]silyl
Synonyms: dimethyl[(trimethylsilyl)oxy]silyl
Molecular Weight (g/mol): 147.34
Molecular Formula: C5H15OSi2
InChi Key: XUKFPAQLGOOCNJ-UHFFFAOYSA-N
SMILES: C[Si](C)O[Si](C)(C)C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results