accessibility menu, dialog, popup

UserName

H3CNNNNNNH2OHClHClHCl

CAS RN 1435934-25-0

IUPAC Name: 7-[4-(4-methylpiperazin-1-yl)cyclohexyl]-5-(4-phenoxyphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine trihydrochloride
Synonyms: 7-[4-(4-methylpiperazin-1-yl)cyclohexyl]-5-(4-phenoxyphenyl)pyrrolo[2,3-d]pyrimidin-4-amine trihydrochloride
Molecular Weight (g/mol): 592.01
Molecular Formula: C29H37Cl3N6O
InChi Key: ALRMEQIQFCUAMR-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.CN1CCN(CC1)C1CCC(CC1)N1C=C(C2=C(N)N=CN=C12)C1=CC=C(OC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results