missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ A 419259 trihydrochloride

Catalog No. 391410
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
391410 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 391410 Supplier Tocris Bioscience™ Supplier No. 3914/10

Inhibitor of Src family kinases

Potent Hck inhibitor (IC50 = 0.43 nM). Also inhibits other Src family kinases (SFKs). Suppresses proliferation and induces apoptosis in CML cell lines including CD34+. Also inhibits acute myeloid leukemia (AML) stem cell proliferation in vitro and in vivo.

Chemical Identifiers

CAS 1435934-25-0
Molecular Formula C29H37Cl3N6O

Specifications

Quantity 10 mg
Formula Weight 592.01
Purity >98%
Molecular Weight (g/mol) 592.006
Chemical Name or Material A 419259 trihydrochloride
Recommended Storage Desiccate at Room Temperature