missing translation for 'onlineSavingsMsg'
Learn More

Emtricitabine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111002322
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002322 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002322 Supplier Merck Emd Millipore Supplier No. PHR2120500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 143491-57-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C8H10 FN3O3 S
Synonym Emtricitabine; (-)-2′3′-Dideoxy-5-fluoro-3′-thiacytidine; (2 R,5 S)-4-Amino-5-fluoro-1-[2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2(1 H)-one; FTC
Type Certified Reference Material
Quantity 500 mg
Formula Weight 247.25
Grade Certified Reference Material